Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC6A18 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP159903
Description
SLC6A18 Polyclonal specifically detects SLC6A18 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SLC6A18 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
FLJ31236, Sodium- and chloride-dependent transporter XTRP2, sodium channel-like protein, sodium-dependent neutral amino acid transporter B(0)AT3, solute carrier family 6 (neurotransmitter transporter), member 18, Solute carrier family 6 member 18, solute carrier family 6, member 18, System B(0) neutral amino acid transporter AT3, XTRP2 | |
Rabbit | |
Protein A purified | |
RUO | |
348932 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1 mg/ml | |
Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
Q96N87 | |
SLC6A18 | |
Synthetic peptides corresponding to SLC6A18(solute carrier family 6, member 18) The peptide sequence was selected from the middle region of SLC6A18. Peptide sequence MHLNATWPKRVAQLPLKACLLEDFLDKSASGPGLAFVVFTETDLHMPGAP. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Rat: 100%; Bovine: 92%; Canine: 92%. | |
Human, Mouse, Rat, Pig, Bovine, Canine, Equine | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction