Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC6A3/DAT1 Antibody (CL3123), Novus Biologicals™

Mouse Monoclonal Antibody
$382.00
Specifications
Antigen | SLC6A3/DAT1 |
---|---|
Clone | CL3123 |
Dilution | Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
SLC6A3/DAT1 Monoclonal antibody specifically detects SLC6A3/DAT1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
SLC6A3/DAT1 | |
Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
Q01959 | |
6531 | |
IgG1 | |
Protein A purified |
CL3123 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Neuroscience | |
PBS (pH 7.2), 40% Glycerol | |
DA transporter, DATDAT1Solute carrier family 6 member 3, sodium-dependent dopamine transporter, solute carrier family 6 (neurotransmitter transporter, dopamine), member 3 | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence LHYLFSSFTTELPWNNSWNSPNCSDAHPGDSSGDSSGLNDTFGTTPAAEYFERGVLHLHQSHGIDDLG | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title