Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC6A3/DAT1 Antibody (mAb16), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB337535 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB337535 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB337535 Supplier Novus Biologicals Supplier No. NBP222164MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SLC6A3/DAT1 Monoclonal antibody specifically detects SLC6A3/DAT1 in Mouse, Rat, Human (Negative) samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)

Specifications

Antigen SLC6A3/DAT1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone mAb16
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias DA transporter, DATDAT1Solute carrier family 6 member 3, sodium-dependent dopamine transporter, solute carrier family 6 (neurotransmitter transporter, dopamine), member 3
Host Species Mouse
Immunogen Synthetic peptide corresponding to the N-terminus of rat DAT1. [UniProt# P23977]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6531
Target Species Mouse, Rat, Human (Negative)
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.