Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC6A3/DAT1 Antibody (mAb16), Novus Biologicals™
SDP

Catalog No. NBP222164SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222164SS 0.025 mL
NBP222164 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP222164SS Supplier Novus Biologicals Supplier No. NBP222164SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 12 publications

SLC6A3/DAT1 Monoclonal specifically detects SLC6A3/DAT1 in Human (Negative), Mouse, Rat samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen SLC6A3/DAT1
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone mAb16
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, ELISA, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:250, Immunoprecipitation reported in scientific literature (PMID 20643191), Immunohistochemistry-Paraffin 1:100
Gene Accession No. Q61327, P23977
Gene Alias DA transporter, DATDAT1Solute carrier family 6 member 3, sodium-dependent dopamine transporter, solute carrier family 6 (neurotransmitter transporter, dopamine), member 3
Gene Symbols SLC6A3
Host Species Mouse
Immunogen Synthetic peptide corresponding to the N-terminus of rat DAT1. [UniProt# P23977]
Purification Method Protein G purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6531
Test Specificity Mouse and rat. Little to no human reactivity has been observed.
Target Species Human (Negative), Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.