Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC6A3/DAT1 Antibody, Novus Biologicals™
SDP

Catalog No. NB300255 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300255 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300255 Supplier Novus Biologicals Supplier No. NB300255
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC6A3/DAT1 Polyclonal specifically detects SLC6A3/DAT1 in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen SLC6A3/DAT1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:1000
Formulation 10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative
Gene Accession No. Q01959
Gene Alias DA transporter, DATDAT1Solute carrier family 6 member 3, sodium-dependent dopamine transporter, solute carrier family 6 (neurotransmitter transporter, dopamine), member 3
Gene Symbols SLC6A3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acid residues from the extracellular loop 2 region conjugated to KLH. Accession # Q01959
Molecular Weight of Antigen 80 kDa
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6531
Test Specificity Specific for the ∼88k DAT protein in Western blots.
Target Species Human, Primate
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.