Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC6A4/5-HTTLPR/Serotonin transporter Antibody (632705), PerCP, Novus Biologicals™
SDP

Catalog No. NB304784 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB304784 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB304784 Supplier Novus Biologicals Supplier No. FAB17292C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SLC6A4/5-HTTLPR/Serotonin transporter Monoclonal specifically detects SLC6A4/5-HTTLPR/Serotonin transporter in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen SLC6A4/5-HTTLPR/Serotonin transporter
Applications Flow Cytometry
Classification Monoclonal
Clone 632705
Conjugate PerCP
Dilution Flow Cytometry
Formulation PBS
Gene Alias 5HT transporter, 5-HTT, hSERT, HTT5-hydroxytryptamine transporter, Na+/Cl- dependent serotonin transporter, OCD1, SERT, sodium-dependent serotonin transporter, solute carrier family 6 (neurotransmitter transporter, serotonin), member 4,5-HTTLPR, Solute carrier family 6 member 4,5HTT
Host Species Mouse
Immunogen Human SLC6A4/5-HTTLPR synthetic peptide, Accession # P31645
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6532
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.