Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC6A4/5-HTTLPR/Serotonin transporter Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB360275 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB360275 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB360275 Supplier Novus Biologicals Supplier No. NBP305546MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC6A4/5-HTTLPR/Serotonin transporter Polyclonal antibody specifically detects SLC6A4/5-HTTLPR/Serotonin transporter in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen SLC6A4/5-HTTLPR/Serotonin transporter
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias 5HT transporter, 5-HTT, hSERT, HTT5-hydroxytryptamine transporter, Na+/Cl- dependent serotonin transporter, OCD1, SERT, sodium-dependent serotonin transporter, solute carrier family 6 (neurotransmitter transporter, serotonin), member 4,5-HTTLPR, Solute carrier family 6 member 4,5HTT
Host Species Rabbit
Immunogen KSITPETPTEIPCGDIRLNAV coupled to KLH
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6532
Target Species Human, Mouse, Rat, Bovine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.