Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC7A14 Antibody, Novus Biologicals™
SDP

Catalog No. NB405018 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
25ul
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB405018 25ul
NBP213346 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB405018 Supplier Novus Biologicals Supplier No. NBP21334625UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

SLC7A14 Polyclonal specifically detects SLC7A14 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SLC7A14
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias FLJ37228, KIAA1613Solute carrier family 7 member 14, probable cationic amino acid transporter, solute carrier family 7 (cationic amino acid transporter, y+ system), member 14
Gene Symbols SLC7A14
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the amino acids: ADCEKEACSPVSEGDEFSGPATNTCGAKNLPSLGDNEMLIGKSDKSTYNVNHPNYGTVDMTTGIEADESENIYLIKLKKLIGPHYYTMRIRL
Purification Method Affinity Purified
Quantity 25ul
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57709
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.