Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC7A5/LAT1 Antibody (BU53), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP2046A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP2046A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP2046A70 Supplier Novus Biologicals Supplier No. NBP250465AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SLC7A5/LAT1 Monoclonal specifically detects SLC7A5/LAT1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Flow (Cell Surface).

Specifications

Antigen SLC7A5/LAT1
Applications Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone BU53
Conjugate Alexa Fluor 750
Dilution Flow Cytometry, Immunohistochemistry, Flow (Cell Surface)
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias 4F2 LC, CD98, CD98LC, D16S469EL-type amino acid transporter 1, E16, Integral membrane protein E16, large neutral amino acids transporter 1, large neutral amino acids transporter small subunit 1, LAT1hLAT1, MPE16CD98 light chain, sodium-independent neutral amino acid transporter LAT1,4F2LC, solute carrier family 7 (cationic amino acid transporter, y+ system), member 5,4F2 light chain, Solute carrier family 7 member 5, y+ system cationic amino acid transporter
Gene Symbols SLC7A5
Host Species Mouse
Immunogen Plasma cell line 8226
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 8140
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.