Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC7A5/LAT1 Antibody (EP3-1), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB340479 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB340479 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB340479 Supplier Novus Biologicals Supplier No. NBP320115MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SLC7A5/LAT1 Monoclonal antibody specifically detects SLC7A5/LAT1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Functional Assay

Specifications

Antigen SLC7A5/LAT1
Applications Immunocytochemistry, Immunoprecipitation, Functional Assay
Classification Monoclonal
Clone EP3-1
Conjugate mFluor Violet 500 SE
Dilution Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Functional
Formulation 50mM Sodium Borate
Gene Alias 4F2 LC, CD98, CD98LC, D16S469EL-type amino acid transporter 1, E16, Integral membrane protein E16, large neutral amino acids transporter 1, large neutral amino acids transporter small subunit 1, LAT1hLAT1, MPE16CD98 light chain, sodium-independent neutral amino acid transporter LAT1,4F2LC, solute carrier family 7 (cationic amino acid transporter, y+ system), member 5,4F2 light chain, Solute carrier family 7 member 5, y+ system cationic amino acid transporter
Host Species Mouse
Immunogen Immunised mice and constructed IgG1 Fab phage display libraries from the spleen cells. SLC7A5/LAT1-reactive recombinant Fab fragments were retrieved by epitope peptide panning.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 8140
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.