Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC7A5/LAT1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB1007340.025MG
Description
SLC7A5/LAT1 Polyclonal specifically detects SLC7A5/LAT1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SLC7A5/LAT1 | |
Polyclonal | |
Unconjugated | |
PBS with No Preservative | |
4F2 LC, CD98, CD98LC, D16S469EL-type amino acid transporter 1, E16, Integral membrane protein E16, large neutral amino acids transporter 1, large neutral amino acids transporter small subunit 1, LAT1hLAT1, MPE16CD98 light chain, sodium-independent neutral amino acid transporter LAT1,4F2LC, solute carrier family 7 (cationic amino acid transporter, y+ system), member 5,4F2 light chain, Solute carrier family 7 member 5, y+ system cationic amino acid transporter | |
Rabbit | |
IgG purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1.9 mg/mL | |
Immunohistochemistry 5 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml | |
Q01650 | |
SLC7A5 | |
KLH conjugated synthetic peptide (RHRKPEL) representing aa 419-425 of Human LAT1. | |
0.025 mg | |
Cancer | |
8140 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction