Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC7A6 Antibody, Novus Biologicals™
SDP

Catalog No. NBP275086 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP275086 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP275086 Supplier Novus Biologicals Supplier No. NBP275086
Add to Cart
Add to Cart

Goat Polyclonal Antibody

SLC7A6 Polyclonal specifically detects SLC7A6 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.

Specifications

Antigen SLC7A6
Applications Western Blot, Immunohistochemistry (Paraffin), ELISA
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 0.3 - 1 ug/mL, Immunohistochemistry-Paraffin 5 ug/mL, Peptide ELISA Detection limit 1:32000
Formulation TBS (pH 7.3), 0.5% BSA with 0.02% Sodium Azide
Gene Alias Y+L amino acid transporter 2, LAT-2, LAT3, solute carrier family 7 (amino acid transporter light chain, y+L system), member 6, y+LAT-2
Gene Symbols SLC7A6
Host Species Goat
Immunogen Peptide with sequence C-ELDVAEEKKDERKTD, from the C Terminus of the protein sequence according to NP_003974.3.
Molecular Weight of Antigen 56.8 kDa
Purification Method Antigen Affinity-purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 9057.0
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.