Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC7A8 Antibody (OTI7D6) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP274215 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274215 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274215 Supplier Novus Biologicals Supplier No. NBP274215
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SLC7A8 Monoclonal specifically detects SLC7A8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen SLC7A8
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI7D6
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:200-1:1000, Immunohistochemistry 1:50
Formulation Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
Gene Alias hLAT2, integral membrane protein E16H, large neutral amino acids transporter small subunit 2, LAT2L-type amino acid transporter 2, LPI-PC1, solute carrier family 7 (amino acid transporter, L-type), member 8, Solute carrier family 7 member 8, y+ system), member 8
Gene Symbols SLC7A8
Host Species Mouse
Immunogen Full-length protein expressed in 293T cell transfected with human SLC7A8 expression vector
Molecular Weight of Antigen 58.4 kDa
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 23428
Reconstitution Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.