Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLCO1C1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.00 - $487.50

Specifications

Antigen SLCO1C1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP15964220
SDP
View Documents
Novus Biologicals
NBP15964220UL
20 μL
Each for $206.00
Only null left
Add to Cart
 
NBP159642
SDP
View Documents
Novus Biologicals
NBP159642
100 μL
Each for $487.50
Only null left
Add to Cart
 
Description

Description

SLCO1C1 Polyclonal specifically detects SLCO1C1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

SLCO1C1
Polyclonal
Rabbit
Q9NYB5
53919
IgG
79 kDa
Western Blot
Unconjugated
RUO
OATP1, OATP14Thyroxine transporter, OATP1C1Solute carrier family 21 member 14, OATPFOAT-RP-5, OATP-FOrganic anion-transporting polypeptide 14, OATPRP5, Organic anion transporter F, Organic anion transporter polypeptide-related protein 5, organic anion transporting polypeptide 14, SLC21A14OATP-14, solute carrier family 21 (organic anion transporter), member 14, solute carrier organic anion transporter family member 1C1, solute carrier organic anion transporter family, member 1C1
SLCO1C1
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.