Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLCO2B1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP180979 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
25ul
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP180979 0.1 mL
NB434008 25ul
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP180979 Supplier Novus Biologicals Supplier No. NBP180979
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLCO2B1 Polyclonal specifically detects SLCO2B1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SLCO2B1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. O94956
Gene Alias OATP2B1OATP-RP2, OATPBDKFZp686E0517, OATP-BKIAA0880, OATPRP2, Organic anion transporter B, Organic anion transporter polypeptide-related protein 2, SLC21A9solute carrier organic anion transporter family member 2B1, solute carrier family 21 (organic anion transporter), member 9, Solute carrier family 21 member 9, solute carrier organic anion transporter family, member 2B1
Gene Symbols SLCO2B1
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:GITHQTSAHPGLELSPSCMEACSCPLDGFNPVCDPSTRVEYITPCHAGCSSWVVQDALDNSQVFYTNCSCVVEGNPVLAGSCDSTCSHLVVPF
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11309
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.