Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLCO3A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB094581 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB094581 20 μL
NB094580 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB094581 Supplier Novus Biologicals Supplier No. NBP30503720UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLCO3A1 Polyclonal specifically detects SLCO3A1 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen SLCO3A1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias FLJ40478, OATP3A1OATP-RP3, OATP-DOATPRP3, OATPDSLC21A11, Organic anion transporter polypeptide-related protein 3, Organic anion-transporting polypeptide D, PGE1 transporter, Sodium-independent organic anion transporter D, solute carrier family 21 (organic anion transporter), member 11, Solute carrier family 21 member 11, solute carrier organic anion transporter family member 3A1, solute carrier organic anion transporter family, member 3A1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 440-540 of human SLCO3A1 (NP_037404.2).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28232
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.