Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLCO5A1 Antibody, Novus Biologicals™
SDP

Catalog No. NB433419 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB433419 25 μL
NBP182498 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB433419 Supplier Novus Biologicals Supplier No. NBP18249825UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

SLCO5A1 Polyclonal specifically detects SLCO5A1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SLCO5A1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q9H2Y9
Gene Alias FLJ39560, member 15, OATP5A1, OATPRP4, organic anion transporter polypeptide-related protein 4, SLC21A15, solute carrier organic anion transporter family, member 5A1
Gene Symbols SLCO5A1
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:SVDAVSDDDVLKEKSNNSEQADKKVSSMGFGKDVRDLPRAAVRILSNM
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81796
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.