Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLCO6A1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen SLCO6A1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP159898
SDP
View Documents
Novus Biologicals
NBP159898
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

SLCO6A1 Polyclonal specifically detects SLCO6A1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

SLCO6A1
Polyclonal
Purified
RUO
Q86UG4
133482
Synthetic peptides corresponding to SLCO6A1(solute carrier organic anion transporter family, member 6A1) The peptide sequence was selected from the N terminal of SLCO6A1. Peptide sequence CCNNIRCFMIFYCILLICQGVVFGLIDVSIGDFQKEYQLKTIEKLALEKS.
Primary
Western Blot
Unconjugated
Rabbit
Human
Cancer/testis antigen 48, CT48Organic anion-transporting polypeptide I, Gonad-specific transporter, GSTOATP-I, MGC26949, OATP6A1Solute carrier family 21 member 19, OATPY, Organic anion-transporting polypeptide 6A1, SLC21A19, solute carrier organic anion transporter family member 6A1, solute carrier organic anion transporter family, member 6A1, testis-specific organic anion transporter
SLCO6A1
IgG
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.