Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLFN12 Antibody, Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB207290 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB207290 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB207290 Supplier Novus Biologicals Supplier No. NBP176252AF647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLFN12 Polyclonal antibody specifically detects SLFN12 in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen SLFN12
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias schlafen family member 12
Host Species Rabbit
Immunogen Antibody was raised against a 12 amino acid synthetic peptide from near the carboxy terminus human SLFN12. The immunogen is located within the last 50 amino acids of SLFN12. Amino Acid Squence: FQKNDKKMFKS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 55106
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.