Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLFN14 Antibody, Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB190462 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB190462 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB190462 Supplier Novus Biologicals Supplier No. NBP282010AF594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLFN14 Polyclonal antibody specifically detects SLFN14 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen SLFN14
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias schlafen family member 14
Host Species Rabbit
Immunogen SLFN14 antibody was raised against a 16 amino acid synthetic peptide from near the carboxy terminus human SLFN14. The immunogen is located within amino acids 730 - 780 of SLFN14. Amino Acid Squence: KVMKEEMKRIKENPPS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 342618
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.