Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLFNL1 Antibody (OTI1G2), FITC, Novus Biologicals™
SDP

Catalog No. NBP274223F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274223F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274223F Supplier Novus Biologicals Supplier No. NBP274223F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SLFNL1 Monoclonal specifically detects SLFNL1 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen SLFNL1
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI1G2
Conjugate FITC
Dilution Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias FLJ23878, MGC43873, schlafen-like 1, schlafen-like protein 1
Gene Symbols SLFNL1
Host Species Mouse
Immunogen Full length recombinant protein of human SLFNL1 (NP_659427) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 200172.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.