Learn More
United Scientific™ Slide Tray, Cardboard, 5 Pk.
SDP
Catalog No. p-200135819 Shop All United Scientific Supplies, Inc Products
Click to view available options
Slide Capacity:
20
30
Protects your slides while holding and transferring them.
Designed as a convenient solution for holding and transferring microscope slides. Fold-over cover helps protect the slides. The flat surface area on the inside of the cover is ideal for labeling or writing any necessary information. Recessed compartments protect the slides when the cover is closed. Thumb cuts on the sides allow you to easily remove each slide. Accommodates microscope slides in two sizes: 75.0 mm L x 25.0 mm W x 0.8 - 1.2 mm D and 76.0 mm L x 26.0 mm W x 0.8 - 1.2 mm D
  • Recessed compartments to protect slides when closed
  • Heavy duty cardboard
  • Thumb cuts for easy removal of slides
  • Flat writing surface for specimen identification
Specifications
Type Slide Storage Tray
Slide Capacity 20
Autoclavable Not Autoclavable
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.