Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLIRP Antibody, DyLight 350, Novus Biologicals™
SDP

Catalog No. N11037258UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N11037258UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N11037258UV Supplier Novus Biologicals Supplier No. NB11037258UV
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLIRP Polyclonal specifically detects SLIRP in Human, Mouse, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen SLIRP
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 350
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias chromosome 14 open reading frame 156, DC50, PD04872, SLIRP, SRA stem-loop-interacting RNA-binding protein, mitochondrial
Gene Symbols SLIRP
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal region of human SLIRP protein. [Entrez-ProtNo. AAX58600]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 81892
Target Species Human, Mouse, Primate
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.