Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLIRP Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB339368 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB339368 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB339368 Supplier Novus Biologicals Supplier No. NB11037258MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLIRP Polyclonal antibody specifically detects SLIRP in Human, Mouse, Primate samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen SLIRP
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias chromosome 14 open reading frame 156, DC50, PD04872, SLIRP, SRA stem-loop-interacting RNA-binding protein, mitochondrial
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal region of human SLIRP protein. [Entrez-Prot# AAX58600]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Chromatin Research, DNA replication Transcription Translation and Splicing, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 81892
Target Species Human, Mouse, Primate
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.