Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Slit1 Antibody, Novus Biologicals™
SDP

Catalog No. NB600880 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600880 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600880 Supplier Novus Biologicals Supplier No. NB600880
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Slit1 Polyclonal specifically detects Slit1 in Mouse, Rat samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Slit1
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1.2 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA 1:10000-1:50000
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. Q80TR4, Q80TR4, Q80TR4, Q80TR4
Gene Alias KIAA0813, MEGF4slit (Drosophila) homolog 1, MGC164811, Multiple EGF-like domains protein 4, Multiple epidermal growth factor-like domains protein 4, SLIL1, slit homolog 1 (Drosophila), slit homolog 1 protein, Slit-1slit1, SLIT3
Gene Symbols SLIT1
Host Species Rabbit
Immunogen This affinity purified Slit1 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region near aa 485-515 of mouse Slit1 protein. (Uniprot: Q80TR4)
Molecular Weight of Antigen 167.42 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Growth and Development, Neuronal Cell Markers, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6585
Test Specificity SLIT1
Target Species Mouse, Rat
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.