Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Slit2 Antibody (710305), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB367991 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB367991 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB367991 Supplier Novus Biologicals Supplier No. FAB5444J
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Slit2 Monoclonal specifically detects Slit2 in Human, Mouse samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen Slit2
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 710305
Conjugate Janelia Fluor 646
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias FLJ14420, slit (Drosophila) homolog 2, slit homolog 2 (Drosophila), slit homolog 2 protein, Slit-2SLIL3
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Slit2, Gln26-Gln900, Accession # Q9R1B9
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Growth and Development, Neuronal Cell Markers, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9353
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.