Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLITRK1 Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB083169 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB083169 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB083169 Supplier Novus Biologicals Supplier No. NBP299232C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLITRK1 Polyclonal specifically detects SLITRK1 in Human samples. It is validated for ELISA.

Specifications

Antigen SLITRK1
Applications ELISA
Classification Polyclonal
Conjugate DyLight 650
Formulation 50 mM Sodium Borate
Gene Alias FLJ54428, KIAA1910KIAA0918, leucine rich repeat containing 12, Leucine-rich repeat-containing protein 12, LRRC12, RP11-395N17.1, SLIT and NTRK-like family, member 1, SLIT and NTRK-like protein 1, slit and trk like gene 1, TTM
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant SLITRK1 (NP_443142.1; Met 1-Ser 616).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 114798
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.