Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLITRK6 Antibody, DyLight 594, Novus Biologicals™
SDP

Catalog No. NB091787 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB091787 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB091787 Supplier Novus Biologicals Supplier No. NBP300225DL594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLITRK6 Polyclonal specifically detects SLITRK6 in Human samples. It is validated for ELISA.

Specifications

Antigen SLITRK6
Applications ELISA
Classification Polyclonal
Conjugate DyLight 594
Formulation 50 mM Sodium Borate
Gene Alias FLJ22774, MGC119595, MGC119596, MGC119597,4832410J21Rik, SLIT and NTRK-like family, member 6, SLIT and NTRK-like protein 6, slit and trk like gene 6
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant SLITRK6 (Q9H5Y7; Met1-Ser608).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84189
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.