Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLK Antibody, Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB056076 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB056076 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB056076 Supplier Novus Biologicals Supplier No. NBP298859AF647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLK Polyclonal specifically detects SLK in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SLK
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias bA16H23.1, CTCL tumor antigen se20-9, EC 2.7.11, EC 2.7.11.1, hSLK, KIAA0204LOSK, Long Ste20-like Kinase, se20-9, Serine/threonine-protein kinase 2, SNF1 (sucrose nonfermenting, yeast, homolog)-like kinase, SNF1 sucrosenonfermenting like kinase, SNF1 (sucrose nonfermenting, yeast, homolog)-like kinase, SNF1 sucrosenonfermenting like kinase (yeast), SNF1 sucrose nonfermenting like kinase, STE20-like kinaseSTE20-like kinase (yeast), STE20-like serine/threonine-protein kinase, STE20-related kinase, Ste20-related serine/threonine kinase, STE20-related serine/threonine-protein kinase, STK2MGC133067
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human SLK.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9748
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.