Learn More
Description
Specifications
Specifications
| Antigen | SLK |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000 |
| Formulation | PBS (pH 7.0) and 20% Glycerol with 0.01% Thimerosal |
| Gene Accession No. | Q9H2G2 |
| Gene Alias | bA16H23.1, CTCL tumor antigen se20-9, EC 2.7.11, EC 2.7.11.1, hSLK, KIAA0204LOSK, Long Ste20-like Kinase, se20-9, Serine/threonine-protein kinase 2, SNF1 (sucrose nonfermenting, yeast, homolog)-like kinase, SNF1 sucrosenonfermenting like kinase, SNF1 (sucrose nonfermenting, yeast, homolog)-like kinase, SNF1 sucrosenonfermenting like kinase (yeast), SNF1 sucrose nonfermenting like kinase, STE20-like kinaseSTE20-like kinase (yeast), STE20-like serine/threonine-protein kinase, STE20-related kinase, Ste20-related serine/threonine kinase, STE20-related serine/threonine-protein kinase, STK2MGC133067 |
| Gene Symbols | SLK |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
