Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Slow Skeletal Muscle Myosin Heavy Chain Antibody (M14), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NBP50297646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP50297646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP50297646 Supplier Novus Biologicals Supplier No. NBP250297JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Slow Skeletal Muscle Myosin Heavy Chain Monoclonal specifically detects Slow Skeletal Muscle Myosin Heavy Chain in Human, Mouse, Rat, Chicken samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Slow Skeletal Muscle Myosin Heavy Chain
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone M14
Conjugate Janelia Fluor 646
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Host Species Mouse
Immunogen purified human skeletal muscle myosin
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cardiovascular Biology, Cellular Signaling, Developmental Biology, Stem Cells
Primary or Secondary Primary
Test Specificity Specific for slow skeletal muscle myosin heavy chain (SM1).
Target Species Chicken, Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.