Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLP-76/LCP2 Antibody (SLP-76/03), FITC, Novus Biologicals™
SDP

Catalog No. NB500537F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500537F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500537F Supplier Novus Biologicals Supplier No. NB500537F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SLP-76/LCP2 Monoclonal specifically detects SLP-76/LCP2 in Human, Mouse, Porcine samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SLP-76/LCP2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SLP-76/03
Conjugate FITC
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS with 0.05% Sodium Azide
Gene Alias lymphocyte cytosolic protein 2, lymphocyte cytosolic protein 2 (SH2 domain containing leukocyte protein of76kDa), lymphocyte cytosolic protein 2 (SH2 domain-containing leukocyte protein of76kD), SH2 domain-containing leukocyte protein of 76 kDa, SH2 domain-containing leukocyte protein of 76kD, SLP-76, SLP-76 tyrosine phosphoprotein, SLP7676 kDa tyrosine phosphoprotein
Gene Symbols LCP2
Host Species Mouse
Immunogen Bacterially expressed fusion protein representing amino acids 216-434 of human SLP76 with histidine tag
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3937
Test Specificity The polyclonal antibody reacts with SLP76, a 76kDa cytosolic adaptor protein that is involved in signaling of various hematopoietic cells, such as T cells, mast cells or neutrophils; in B cells, however, it is replaced by SLP65.
Target Species Human, Mouse, Pig
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.