Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ SLU-PP-332

Click to view available options
Quantity:
10 mg
50 mg
Description
SLU-PP-332 is a potent and selective pan estrogen related receptor (ERR) agonist (EC50 values are 98 nM, 230 nM and 430 nM for ERRα, ERRβ and ERRγ respectively). In a mouse model of metabolic syndrome SLU-PP-332 increases energy expenditure and fatty acid oxidation, decreases fat mass, and improves insulin sensitivity. Also increases mitochondrial respiration in vitro.
Specifications
Specifications
Chemical Name or Material | (E)-4-Hydroxy-N'-(naphthalen-2-ylmethylene)benzohydrazide |
CAS | 303760-60-3 |
Quantity | 50 mg |
Target | Estrogen and Related Receptor Agonists |
Molecular Formula | C18H14N2O2 |
Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction