Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Slug Antibody (666633), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB374716 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB374716 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB374716 Supplier Novus Biologicals Supplier No. FAB7408Y
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Slug Monoclonal antibody specifically detects Slug in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOF

Specifications

Antigen Slug
Applications Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone 666633
Conjugate DyLight 680
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias MGC10182, Neural crest transcription factor Slug, Protein snail homolog 2, slug homolog, zinc finger protein (chicken), SLUGH, SLUGzinc finger protein, snail 2, snail homolog 2 (Drosophila), WS2D, zinc finger protein SNAI2
Host Species Mouse
Immunogen E. coli-derived recombinant human Slug, Met1-His268, Accession # O43623
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuronal Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6591
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.