Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Slug Antibody (OTI1A6), FITC, Novus Biologicals™
SDP

Catalog No. NBP274229F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274229F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274229F Supplier Novus Biologicals Supplier No. NBP274229F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Slug Monoclonal specifically detects Slug in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry.

Specifications

Antigen Slug
Applications Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI1A6
Conjugate FITC
Dilution Flow Cytometry, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias MGC10182, Neural crest transcription factor Slug, Protein snail homolog 2, slug homolog, zinc finger protein (chicken), SLUGH, SLUGzinc finger protein, snail 2, snail homolog 2 (Drosophila), WS2D, zinc finger protein SNAI2
Gene Symbols SNAI2
Host Species Mouse
Immunogen Full length recombinant protein of human Slug (NP_003059) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Neuronal Cell Markers, Neuronal Stem Cell Markers, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 6591.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.