Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals SM buffer, pH 7.5

Catalog No. p-7042107
Click to view available options
Quantity:
100 mL
250 mL

SM buffer is mainly used in molecular biology lab. It is used for routine manipulation of phage suspensions. The gelatin in SM buffer stabilizes lambda phage particles during storage. is mainly used in molecular biology lab.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
SM buffer is mainly used in molecular biology lab. It is used for routine manipulation of phage suspensions. The gelatin in SM buffer stabilizes lambda phage particles during storage. is mainly used in molecular biology lab.

Solubility
Not miscible in water.

Notes
50mM Tris-HCl, 8mM magnesium sulfate, 100mM sodium chloride, and 0.01% gelatin, pH7.5
TRUSTED_SUSTAINABILITY

Specifications

pH 7.5
Quantity 100 mL
Content And Storage Ambient Temperatures
Form Liquid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.