Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMAC/Diablo Antibody (006), HRP, Novus Biologicals™
SDP

Catalog No. NB088194 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB088194 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB088194 Supplier Novus Biologicals Supplier No. NBP289251H
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

SMAC/Diablo Monoclonal specifically detects SMAC/Diablo in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen SMAC/Diablo
Applications Western Blot, ELISA
Classification Monoclonal
Clone 6
Conjugate HRP
Formulation PBS
Gene Alias 0610041G12Rik, diablo, IAP-binding mitochondrial protein, Direct IAP-binding protein with low pI, mitochondrial Smac protein, Second mitochondria-derived activator of caspase, Smac, SMACmitochondrial
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human SMAC/Diablo (NP_063940.1; Ala 56-Asp 239).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Cellular Markers, Mitochondrial Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 56616
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.