Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMAC/Diablo Antibody, Novus Biologicals™
SDP

Catalog No. NB10056311 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056311 0.1 mg
NB10056311T 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056311 Supplier Novus Biologicals Supplier No. NB10056311
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 16 publications

SMAC/Diablo Polyclonal specifically detects SMAC/Diablo in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen SMAC/Diablo
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-2 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:500. Use reported in scientific literature (Hu et al (2004)), Immunohistochemistry-Paraffin 10 ug/ml
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. Q9NR28
Gene Alias 0610041G12Rik, diablo, IAP-binding mitochondrial protein, Direct IAP-binding protein with low pI, mitochondrial Smac protein, Second mitochondria-derived activator of caspase, Smac, SMACmitochondrial
Gene Symbols DIABLO
Host Species Rabbit
Immunogen This rabbit polyclonal antibody was developed against two synthetic peptides corresponding to amino acids 199 to 212 and amino acids 226 to 239 of human Smac (Genbank accession no. AAF87716).
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56616
Test Specificity Smac
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.