Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZIC1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB600488 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600488 0.1 mL
NB600488SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB600488 Supplier Novus Biologicals Supplier No. NB600488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 4 publications

ZIC1 Polyclonal specifically detects ZIC1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen ZIC1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:400, Immunohistochemistry-Frozen reported in scientific literature (PMID 23862012)
Formulation PBS and 30% Glycerol with 0.01% Sodium Azide
Gene Accession No. Q15915
Gene Alias Zic family member 1 (odd-paired homolog, Drosophila), ZICZic family member 1 (odd-paired Drosophila homolog), Zinc finger protein 201, Zinc finger protein of the cerebellum 1, ZNF201zinc finger protein ZIC 1
Gene Symbols ZIC1
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human Zic1 protein (between residues 20-75) [UniProt Q15915]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Growth and Development, Neuronal Cell Markers, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7545
Test Specificity Human and mouse. Predicted to react with rat based on 100% sequence homology.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.