Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Smad2, p Ser465, p Ser467 Antibody, Novus Biologicals™
SDP

Catalog No. NBP244217 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP244217 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP244217 Supplier Novus Biologicals Supplier No. NBP244217
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Smad2/3 in Human samples, and it is validated for ELISA, Western Blotting

Smad2 Polyclonal specifically detects Smad2 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Smad2 (p Ser465, p Ser467)
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.94 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:3000, ELISA 1:10000
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. Q15796
Gene Alias hMAD-2, hSMAD2, JV18, JV18-1SMAD, mothers against DPP homolog 2 (Drosophila), MAD homolog 2, Mad protein homolog, Mad, mothers against decapentaplegic homolog 2, MADH2MGC34440, MADR2MGC22139, Mad-related protein 2, mother against DPP homolog 2, mothers against decapentaplegic homolog 2, Mothers against DPP homolog 2, Sma- and Mad-related protein 2, SMAD 2, SMAD family member 2MAD, mothers against decapentaplegic homolog 2 (Drosophila), SMAD, mothers against DPP homolog 2, Smad2
Gene Symbols SMAD2
Host Species Rabbit
Immunogen Smad2 [p Ser465, p Ser467] Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to a C-terminal of human Smad2 protein. (Uniprot: Q15796)
Molecular Weight of Antigen 52.31 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Stem Cell Signaling Pathway, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 4087
Test Specificity Anti-Smad2 pS465 pS467 antibody is directed against the phosphorylated form of human Smad2 protein at the pS465 and pS467 residues. The resultant affinity purified antibody was then cross adsorbed against the non-phosphorylated form of the immunizing peptide. Reactivity occurs against human Smad2. Reactivity with non-phosphorylated human Smad2 is minimal by ELISA and western blot. A BLAST analysis was used to suggest cross-reactivity with Smad2 protein from human, mouse, rat, orangutan, and bovine based on 100% homology with the immunizing sequence. Reactivity against homologues from other sources is not known.
Target Species Human, Mouse, Rat, Bovine, Orangutan
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.