Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Smad2, p Ser467 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP254771 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
0.1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP254771 0.1 mg
NBP25477105 0.05 mg
NB413813 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP254771 Supplier Novus Biologicals Supplier No. NBP254771
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Smad2 Polyclonal specifically detects Smad2 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Smad2 (p Ser467)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000
Formulation Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl and 50% glycerol with 0.02% Sodium Azide
Gene Alias hMAD-2, hSMAD2, JV18, JV18-1SMAD, mothers against DPP homolog 2 (Drosophila), MAD homolog 2, Mad protein homolog, Mad, mothers against decapentaplegic homolog 2, MADH2MGC34440, MADR2MGC22139, Mad-related protein 2, mother against DPP homolog 2, mothers against decapentaplegic homolog 2, Mothers against DPP homolog 2, pSMAD, Sma- and Mad-related protein 2, SMAD 2, SMAD family member 2MAD, mothers against decapentaplegic homolog 2 (Drosophila), SMAD, mothers against DPP homolog 2, Smad2
Gene Symbols SMAD2
Host Species Rabbit
Immunogen Smad2 (Phospho-Ser467) antibody was raised against a peptide sequence around phosphorylation site of serine 467 (C-S-S-M-S (p) ) derived from Human Smad2.
Molecular Weight of Antigen 60 kDa
Purification Method Epitope affinity purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Metastasis, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 4087
Test Specificity This antibody detects endogenous level of Smad2 only when phosphorylated at serine 467.
Target Species Human, Mouse, Rat
Content And Storage Store at -20 degrees C. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.