Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Smad2 (p Ser250) Rabbit anti-Human, Mouse, Rat, Clone: SD207-1, Novus Biologicals™
SDP

Catalog No. NBP267761 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267761 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267761 Supplier Novus Biologicals Supplier No. NBP267761

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Monoclonal Antibody

Smad2 Monoclonal specifically detects Smad2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen Smad2 (p Ser250)
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone SD207-1
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:50-1:100
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias hMAD-2, hSMAD2, JV18, JV18-1SMAD, mothers against DPP homolog 2 (Drosophila), MAD homolog 2, Mad protein homolog, Mad, mothers against decapentaplegic homolog 2, MADH2MGC34440, MADR2MGC22139, Mad-related protein 2, mother against DPP homolog 2, mothers against decapentaplegic homolog 2, Mothers against DPP homolog 2, Sma- and Mad-related protein 2, SMAD 2, SMAD family member 2MAD, mothers against decapentaplegic homolog 2 (Drosophila), SMAD, mothers against DPP homolog 2, Smad2
Gene Symbols SMAD2
Host Species Rabbit
Immunogen Synthetic phospho-peptide corresponding to residues surrounding Ser250 of Human Smad2 aa 221-270/ 467. (SwissProt: Q15796 Human; SwissProt: Q62432 Mouse; SwissProt: O70436 Rat)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Cancer, Cell Cycle and Replication, Metastasis, Phospho Specific, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 4087
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.