Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Smad4 Antibody (253343), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB304586 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB304586 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB304586 Supplier Novus Biologicals Supplier No. FAB2097N

Mouse Monoclonal Antibody

Smad4 Monoclonal antibody specifically detects Smad4 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF

Specifications

Antigen Smad4
Applications Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 253343
Conjugate Alexa Fluor 700
Dilution Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias DPC4SMAD, mothers against DPP homolog 4 (Drosophila), hSMAD4, JIP, MAD homolog 4, MADH4mothers against decapentaplegic homolog 4, mothers against decapentaplegic homolog 4, mothers against decapentaplegic, Drosophila, homolog of, 4, SMAD 4, SMAD family member 4MAD, mothers against decapentaplegic homolog 4 (Drosophila), SMAD, mothers against DPP homolog 4, Smad4
Host Species Mouse
Immunogen E. coli-derived recombinant human Smad4, Pro139-Asp332, Accession # Q13485
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction, Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 4089
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.