Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Smad4 Antibody (SMAD4/2440), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB387552 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB387552 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB387552 Supplier Novus Biologicals Supplier No. NBP308320JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Smad4 Monoclonal antibody specifically detects Smad4 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen Smad4
Applications Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone SMAD4/2440
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias DPC4SMAD, mothers against DPP homolog 4 (Drosophila), hSMAD4, JIP, MAD homolog 4, MADH4mothers against decapentaplegic homolog 4, mothers against decapentaplegic homolog 4, mothers against decapentaplegic, Drosophila, homolog of, 4, SMAD 4, SMAD family member 4MAD, mothers against decapentaplegic homolog 4 (Drosophila), SMAD, mothers against DPP homolog 4, Smad4
Host Species Mouse
Immunogen Recombinant full-length human Smad4 protein (Uniprot: Q13485 )
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signal Transduction, Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 4089
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.