Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Smad5 Antibody (3H9), Novus Biologicals™
SDP

Catalog No. NBP237648 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP237648 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP237648 Supplier Novus Biologicals Supplier No. NBP237648
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Smad5 Monoclonal specifically detects Smad5 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Smad5
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 3H9
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000
Formulation Ascites with 0.03% Sodium Azide
Gene Alias hSmad5, JV5-1DKFZp781O1323, MAD homolog 5, MAD, mothers against decapentaplegic homolog 5 (Drosophila), MADH5mothers against decapentaplegic homolog 5, mothers against decapentaplegic homolog 5, mothers against decapentaplegic, drosophila, homolog of, 5, Mothers against DPP homolog 5, SMA- and MAD-related protein 5, SMAD 5, SMAD family member 5DKFZp781C1895, SMAD, mothers against DPP homolog 5, SMAD, mothers against DPP homolog 5 (Drosophila), Smad5
Gene Symbols SMAD5
Host Species Mouse
Immunogen Purified recombinant fragment of human SMAD5 expressed in E. Coli.
Molecular Weight of Antigen 52 kDa
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4090
Target Species Human, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.