Learn More
Description
Specifications
Specifications
| Antigen | SMAD6 |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Simple Western, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.5 - 2 ug/mL. Use reported by customer review, Simple Western 1:50, Immunohistochemistry 5 - 10 ug/mL, Immunocytochemistry/Immunofluorescence 1 - 5 ug/mL. Use reported in scientific literature (PMID 20032053), Immunohistochemistry-Paraffin 5 - 10 ug/mL. Use reported in scientific literature (PMID 20032053) |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Accession No. | O43541 |
| Gene Alias | hSMAD6, HsT17432, MAD homolog 6, MADH6SMAD, mothers against DPP homolog 6 (Drosophila), MADH7, mothers against decapentaplegic homolog 6, Mothers against decapentaplegic, drosophila, homolog of, 6, Mothers against DPP homolog 6, SMAD 6, SMAD family member 6MAD, mothers against decapentaplegic homolog 6 (Drosophila), SMAD, mothers against DPP homolog 6, Smad6 |
| Gene Symbols | SMAD6 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
