Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMAD6 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056440T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056440T 0.025 mg
NB10056440 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056440T Supplier Novus Biologicals Supplier No. NB10056440SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 8 publications

SMAD6 Polyclonal specifically detects SMAD6 in Human, Monkey, Mouse, Ovine, Primate, Rat, Sheep samples. It is validated for IF/IHC, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Simple Western, Western Blot.

Specifications

Antigen SMAD6
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Simple Western, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 2 ug/mL. Use reported by customer review, Simple Western 1:50, Immunohistochemistry 5 - 10 ug/mL, Immunocytochemistry/Immunofluorescence 1 - 5 ug/mL. Use reported in scientific literature (PMID 20032053), Immunohistochemistry-Paraffin 5 - 10 ug/mL. Use reported in scientific literature (PMID 20032053)
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. O43541
Gene Alias hSMAD6, HsT17432, MAD homolog 6, MADH6SMAD, mothers against DPP homolog 6 (Drosophila), MADH7, mothers against decapentaplegic homolog 6, Mothers against decapentaplegic, drosophila, homolog of, 6, Mothers against DPP homolog 6, SMAD 6, SMAD family member 6MAD, mothers against decapentaplegic homolog 6 (Drosophila), SMAD, mothers against DPP homolog 6, Smad6
Gene Symbols SMAD6
Host Species Rabbit
Immunogen This antibody was developed against 2 synthetic peptides found between amino acids 30-150 and 300-400 of human SMAD6 (GenBank Accession No. AAH12986.1). These peptide sequences are specific to SMAD6, and have high homology to SMAD6 in multiple species.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4091
Test Specificity A 52 kDa band is observed.
Target Species Human, Mouse, Monkey, Mouse, Ovine, Sheep, Primate, Rat, Sheep
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.