Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMAD6 Antibody, Novus Biologicals™
SDP

Catalog No. NB035932 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB035932 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB035932 Supplier Novus Biologicals Supplier No. NBP288310
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SMAD6 Polyclonal specifically detects SMAD6 in Human samples. It is validated for Western Blot.

Specifications

Antigen SMAD6
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose
Gene Alias hSMAD6, HsT17432, MAD homolog 6, MADH6SMAD, mothers against DPP homolog 6 (Drosophila), MADH7, mothers against decapentaplegic homolog 6, Mothers against decapentaplegic, drosophila, homolog of, 6, Mothers against DPP homolog 6, SMAD 6, SMAD family member 6MAD, mothers against decapentaplegic homolog 6 (Drosophila), SMAD, mothers against DPP homolog 6, Smad6
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the N-terminal region of Human SMAD6
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 4091
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.