Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Smad7 Antibody, Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB056935 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB056935 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB056935 Supplier Novus Biologicals Supplier No. NBP298971AF488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Smad7 Polyclonal specifically detects Smad7 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Smad7
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias CRCS3, FLJ16482, hSMAD7, MAD homolog 7, MAD homolog 8, MAD, mothers against decapentaplegic homolog 7 (Drosophila), MADH7mothers against decapentaplegic homolog 7, MADH8, Mothers against decapentaplegic homolog 8, Mothers against decapentaplegic, drosophila, homolog of, 7, Mothers against DPP homolog 7, Mothers against DPP homolog 8, SMAD 7, SMAD family member 7MAD (mothers against decapentaplegic, Drosophila) homolog 7, SMAD, mothers against DPP homolog 7, SMAD, mothers against DPP homolog 7 (Drosophila), Smad7
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human Smad7.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4092
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.