Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Smad7 Rabbit anti-Human, Mouse, Sheep, Alexa Fluor 594, Polyclonal, Novus Biologicals™
SDP

Catalog No. NP224710A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP224710A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP224710A94 Supplier Novus Biologicals Supplier No. NBP224710AF594

Rabbit Polyclonal Antibody

Smad7 Polyclonal specifically detects Smad7 in Human, Mouse, Sheep samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Smad7
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 594
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CRCS3, FLJ16482, hSMAD7, MAD homolog 7, MAD homolog 8, MAD, mothers against decapentaplegic homolog 7 (Drosophila), MADH7mothers against decapentaplegic homolog 7, MADH8, Mothers against decapentaplegic homolog 8, Mothers against decapentaplegic, drosophila, homolog of, 7, Mothers against DPP homolog 7, Mothers against DPP homolog 8, SMAD 7, SMAD family member 7MAD (mothers against decapentaplegic, Drosophila) homolog 7, SMAD, mothers against DPP homolog 7, SMAD, mothers against DPP homolog 7 (Drosophila), Smad7
Gene Symbols SMAD7
Host Species Rabbit
Immunogen This antibody was raised against two peptides made from within amino acids 10 and 50 of human SMAD7. This sequence is 100% or highly conserved between human and multiple species.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 4092
Test Specificity This antibody may cross-react with SMAD6 depending on cell or tissue-specific expression.
Target Species Human, Mouse, Sheep
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.